siRNA Details
| Gene Symbol: RNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022453.2 | NP_071898.2 | 3 | 220 | ||
| XM_017004695.1 | XP_016860184.1 | 3 | 220 | |||
| GenBank mRNA | AK023968.1 | - | 3 | 138 | ||
| AK222525.1 | - | 3 | 158 | |||
| AK289501.1 | - | 3 | 164 | |||
| BC015612.2 | - | 3 | 137 | 4811222 | ||
| BI831198.1 | - | 3 | 220 | |||

RNF25
Genomic Map