siRNA Details
| Gene Symbol: AKTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012398.2 | NP_001012398.1 | 7 | 793 | ||
| NM_001308325.1 | NP_001295254.1 | 7 | 764 | |||
| NM_022476.3 | NP_071921.1 | 7 | 764 | |||
| XM_005256095.4 | XP_005256152.1 | 7 | 1113 | |||
| XM_005256096.4 | XP_005256153.1 | 7 | 691 | |||
| XM_005256097.4 | XP_005256154.1 | 7 | 725 | |||
| XM_005256098.4 | XP_005256155.1 | 7 | 689 | |||
| XM_017023564.1 | XP_016879053.1 | 7 | 1113 | |||
| XM_017023565.1 | XP_016879054.1 | 7 | 691 | |||
| XM_017023566.1 | XP_016879055.1 | 7 | 689 | |||
| GenBank mRNA | AK023320.1 | - | 7 | 699 | ||
| AK222743.1 | - | 7 | 696 | |||
| BC001134.2 | - | 7 | 712 | 2967019 | ||
| BC095401.1 | - | 7 | 686 | 30334599 | ||
| CR457308.1 | - | 6 | 536 | |||

AKTIP
Genomic Map