siRNA Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023079.4 | NP_075567.2 | 3 | 630 | ||
| GenBank mRNA | AB025426.1 | - | 2 | 171 | ||
| AK023917.1 | - | 2 | 168 | |||
| AK024030.1 | - | 4 | 485 | |||
| AK091644.1 | - | 1 | 1761 | |||
| AK122770.1 | - | 3 | 528 | |||
| AK130303.1 | - | 4 | 488 | |||
| AK296865.1 | - | 4 | 521 | |||
| BC015169.1 | - | 2 | 157 | |||
| BC015890.1 | - | 3 | 498 | 3915698 | ||
| CR457322.1 | - | 2 | 171 | |||
| EF623992.1 | - | 3 | 495 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172300, AK005524, AK152529, BC027266, AK153328, and others.

UBE2Z
Genomic Map