siRNA Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023079.4 | NP_075567.2 | 7 | 1449 | ||
| GenBank mRNA | AK023917.1 | - | 6 | 987 | ||
| AK024030.1 | - | 8 | 1304 | |||
| AK091644.1 | - | 5 | 2580 | |||
| AK122770.1 | - | 7 | 1347 | |||
| AK130303.1 | - | 8 | 1307 | |||
| AK296865.1 | - | 8 | 1340 | |||
| AL713782.1 | - | 3 | 604 | |||
| BC015169.1 | - | 6 | 976 | |||
| BC015890.1 | - | 7 | 1317 | 3915698 | ||
 Back To Top
 Back To Top
 UBE2Z
UBE2Z Genomic Map
Genomic Map Back To Top
 Back To Top