siRNA Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023079.4 | NP_075567.2 | 7 | 1881 | ||
| GenBank mRNA | AK023917.1 | - | 6 | 1419 | ||
| AK024030.1 | - | 8 | 1736 | |||
| AK091644.1 | - | 5 | 3012 | |||
| AK122770.1 | - | 7 | 1779 | |||
| AK130303.1 | - | 8 | 1739 | |||
| AL713782.1 | - | 3 | 1036 | |||
| BC015169.1 | - | 6 | 1408 | |||
| BC015890.1 | - | 7 | 1749 | 3915698 | ||

UBE2Z
Genomic Map