siRNA Details
| Gene Symbol: ANKRD30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052997.2 | NP_443723.2 | 7 | 1071 | ||
| XM_011519757.2 | XP_011518059.1 | 7 | 1174 | |||
| XM_011519758.2 | XP_011518060.1 | 7 | 1174 | |||
| GenBank mRNA | AF269087.1 | - | 7 | 1071 | ||

ANKRD30A
Genomic Map