siRNA Details
Gene Symbol: OTUD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130901.1 | NP_570971.1 | 4 | 682 | ||
GenBank mRNA | AJ430383.1 | - | 4 | 682 | ||
BC035668.1 | - | 6 | 758 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OTUD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130901.1 | NP_570971.1 | 4 | 682 | ||
GenBank mRNA | AJ430383.1 | - | 4 | 682 | ||
BC035668.1 | - | 6 | 758 |