siRNA Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282607.1 | NP_001269536.1 | 12 | 2716 | ||
| NM_144967.3 | NP_659404.2 | 12 | 2952 | |||
| XM_006724729.1 | XP_006724792.1 | 11 | 2423 | |||
| XM_011531280.1 | XP_011529582.1 | 11 | 2440 | |||
| GenBank mRNA | AK054620.1 | - | 9 | 2312 | ||
| AK128203.1 | - | 12 | 2951 | |||
| BC063790.1 | - | 12 | 2551 | 6196091 | ||
| CD107623.1 | - | 1 | 699 | |||

ARHGAP36
Genomic Map