siRNA Details
| Gene Symbol: GPHB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145171.3 | NP_660154.3 | 3 | 300 | ||
| GenBank mRNA | AF403430.1 | - | 3 | 243 | ||
| AF467770.1 | - | 2 | 243 | |||
| BC069113.1 | - | 3 | 243 | 7216930 | ||
| BC133702.1 | - | 2 | 243 | |||
| BM709443.1 | - | 3 | 300 | |||

GPHB5
Genomic Map