siRNA Details
| Gene Symbol: UBXN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152376.4 | NP_689589.1 | 2 | 2221 | ||
| XM_005245742.3 | XP_005245799.1 | 2 | 2725 | |||
| XM_011540699.2 | XP_011539001.1 | 2 | 2429 | |||
| GenBank mRNA | BC036417.1 | - | 1 | 1631 | ||
| BC047428.1 | - | 2 | 2216 | 5295853 | ||
| BX648631.1 | - | 2 | 2211 | |||

UBXN10
Genomic Map