siRNA Details
| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152489.1 | NP_689702.1 | 2, 3 | 399 | ||
| XM_005270498.4 | XP_005270555.1 | 2, 3 | 1193 | |||
| XM_006710379.3 | XP_006710442.1 | 2, 3 | 334 | |||
| XM_011540764.2 | XP_011539066.1 | 3, 4 | 1134 | |||
| GenBank mRNA | BC029895.1 | - | 2, 3 | 399 | 5168406 | |

UBE2U
Genomic Map