siRNA Details
| Gene Symbol: C3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173824.3 | NP_776185.2 | 2 | 598 | ||
| XM_005264745.4 | XP_005264802.1 | 2 | 627 | |||
| GenBank mRNA | AK297719.1 | - | 2 | 354 | ||
| AK313547.1 | - | 2 | 339 | |||
| AL832398.1 | - | 2 | 598 | |||
| BC024188.2 | - | 2 | 382 | 4824439 | ||
| KU178900.1 | - | 2 | 281 | |||
| KU178901.1 | - | 2 | 281 | |||

C3orf38
Genomic Map