siRNA Details
| Gene Symbol: NME9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178130.3 | NP_835231.1 | 12 | 1013 | ||
| GenBank mRNA | AK094830.1 | - | 12 | 1013 | ||
| AY336746.1 | - | 8 | 703 | |||
| BC107054.2 | - | 8 | 652 | 40005048 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NME9| Gene Symbol: NME9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178130.3 | NP_835231.1 | 12 | 1013 | ||
| GenBank mRNA | AK094830.1 | - | 12 | 1013 | ||
| AY336746.1 | - | 8 | 703 | |||
| BC107054.2 | - | 8 | 652 | 40005048 | ||