siRNA Details
| Gene Symbol: NRBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178564.3 | NP_848659.2 | 18 | 2270 | ||
| XM_017013380.1 | XP_016868869.1 | 17 | 2361 | |||
| XM_017013383.1 | XP_016868872.1 | 17 | 2214 | |||
| GenBank mRNA | AF318376.1 | - | 8 | 1609 | ||
| AK094017.1 | - | 2 | 1255 | |||
| AK308967.1 | - | 4 | 905 | |||
| AL137662.1 | - | 16 | 2199 | |||
| AL834530.1 | - | 11 | 1509 | |||
| BC071605.1 | - | 17 | 2433 | |||

NRBP2
Genomic Map