siRNA Details
| Gene Symbol: NRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198465.3 | NP_940867.2 | 7 | 818 | ||
| XM_006724632.3 | XP_006724695.1 | 7 | 780 | |||
| XM_006724633.3 | XP_006724696.1 | 7 | 780 | |||
| XM_006724634.3 | XP_006724697.1 | 7 | 780 | |||
| XM_011530887.2 | XP_011529189.1 | 7 | 572 | |||
| GenBank mRNA | AK131549.1 | - | 7 | 780 | ||
| BX538345.1 | - | 7 | 820 | |||

NRK
Genomic Map