siRNA Details
Gene Symbol: PSMB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099780.1 | NP_001093250.1 | 1 | 1134 | ||
GenBank mRNA | AK122797.1 | - | 1 | 1134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PSMB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099780.1 | NP_001093250.1 | 1 | 1134 | ||
GenBank mRNA | AK122797.1 | - | 1 | 1134 |