siRNA Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243116.1 | NP_001230045.1 | 3 | 466 | ||
| NM_002503.4 | NP_002494.2 | 3 | 708 | |||
| NR_040515.1 | 3 | 652 | ||||
| XM_006723226.2 | XP_006723289.2 | 3 | 644 | |||
| XM_006723227.3 | XP_006723290.3 | 3 | 607 | |||
| GenBank mRNA | AB209784.1 | - | 3 | 674 | ||
| AK057862.1 | - | 3 | 603 | |||
| AK290569.1 | - | 3 | 659 | |||
| BC007197.1 | - | 1 | 308 | |||
| BC015528.1 | - | 3 | 639 | 3903794 | ||
| BI520169.1 | - | 3 | 466 | |||
| BM449613.1 | - | 3 | 663 | |||
| BQ440907.1 | - | 3 | 440 | |||
| BT006743.1 | - | 3 | 605 | |||

NFKBIB
Genomic Map