siRNA Details
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002314.3 | NP_002305.1 | 2 | 314 | ||
GenBank mRNA | AF134379.1 | - | 2 | 100 | ||
AK290162.1 | - | 3 | 239 | |||
D26309.1 | - | 3 | 195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002314.3 | NP_002305.1 | 2 | 314 | ||
GenBank mRNA | AF134379.1 | - | 2 | 100 | ||
AK290162.1 | - | 3 | 239 | |||
D26309.1 | - | 3 | 195 |