siRNA Details
| Gene Symbol: PSMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002812.4 | NP_002803.2 | 5 | 774 | ||
| GenBank mRNA | AK301771.1 | - | 5 | 785 | ||
| AK304486.1 | - | 5 | 359 | |||
| AK308809.1 | - | 5 | 863 | |||
| BC001164.1 | - | 5 | 525 | 3528096 | ||
| BC065006.1 | - | 5 | 513 | 6055235 | ||
| BQ684248.1 | - | 5 | 537 | |||
| BT019688.1 | - | 5 | 443 | |||
| D38047.1 | - | 5 | 513 | |||
| EU668354.1 | - | 5 | 774 | |||

PSMD8
Genomic Map