siRNA Details
| Gene Symbol: TECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321170.1 | NP_001308099.1 | 12 | 883 | ||
| NM_138501.5 | NP_612510.1 | 11 | 809 | |||
| NR_038103.1 | 11 | 841 | ||||
| NR_038104.1 | 11 | 874 | ||||
| XM_006722945.1 | XP_006723008.1 | 12 | 927 | |||
| XM_011528442.2 | XP_011526744.1 | 11 | 757 | |||
| GenBank mRNA | AF038958.1 | - | 11 | 755 | ||
| AF038959.1 | - | 6 | 238 | |||
| AF222742.1 | - | 11 | 730 | |||
| AK001416.1 | - | 11 | 773 | |||
| AK094087.1 | - | 12 | 850 | |||
| AK299134.1 | - | 12 | 847 | |||
| AK315412.1 | - | 11 | 773 | |||
| BC000174.2 | - | 10 | 669 | |||
| BC002425.1 | - | 11 | 741 | 3163592 | ||
| BC005952.1 | - | 11 | 752 | 4249841 | ||
| BC007801.2 | - | 11 | 759 | 4128702 | ||
| BC013881.2 | - | 11 | 757 | 3928698 | ||
| BT007179.1 | - | 10 | 679 | |||
| CR456892.1 | - | 10 | 679 | |||

TECR
Genomic Map