siRNA Details
| Gene Symbol: APC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000038.5 | NP_000029.2 | 16 | 3510 | ||
| NM_001127510.2 | NP_001120982.1 | 17 | 3618 | |||
| NM_001127511.2 | NP_001120983.2 | 14 | 3584 | |||
| GenBank mRNA | AB210001.1 | - | 17 | 3582 | ||
| AK289567.1 | - | 16 | 3572 | |||
| AK294544.1 | - | 14 | 3568 | |||
| AK309033.1 | - | 3 | 1779 | |||
| M73548.1 | - | 15 | 3463 | |||
| M74088.1 | - | 15 | 3443 | |||

APC
Genomic Map