siRNA Details
| Gene Symbol: B3GNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138706.4 | NP_619651.3 | 2 | 2244 | ||
| GenBank mRNA | AK127544.1 | - | 2 | 1827 | ||
| AK172863.1 | - | 2 | 2169 | |||
| BC025357.1 | - | 1 | 1785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3GNT6| Gene Symbol: B3GNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138706.4 | NP_619651.3 | 2 | 2244 | ||
| GenBank mRNA | AK127544.1 | - | 2 | 1827 | ||
| AK172863.1 | - | 2 | 2169 | |||
| BC025357.1 | - | 1 | 1785 | |||