siRNA Details
| Gene Symbol: TBC1D26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178571.4 | NP_848666.2 | 14, 15 | 1335 | ||
| GenBank mRNA | BC028414.1 | - | 11, 12 | 3573 | ||
| BC047400.1 | - | 14, 15 | 1329 | 5297645 | ||
| Gene Symbol: LOC105369205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524131.2 | XP_011522433.1 | 4 | 3484 | ||

TBC1D26
Genomic Map