siRNA Details
| Gene Symbol: NAT8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016347.2 | NP_057431.2 | 1 | 138 | ||
| NR_132338.1 | 1 | 138 | ||||
| GenBank mRNA | AF185571.1 | - | 1 | 138 | ||
| BC069564.1 | - | 1 | 138 | |||
| BC121101.1 | - | 1 | 142 | |||
| BC121102.1 | - | 1 | 142 | |||

NAT8B
Genomic Map