siRNA Details
| Gene Symbol: TPTE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290224.1 | NP_001277153.1 | 19 | 2278 | ||
| NM_199259.3 | NP_954868.1 | 23 | 2500 | |||
| NM_199260.3 | NP_954869.1 | 22 | 2440 | |||
| NM_199261.3 | NP_954870.2 | 24 | 2554 | |||
| GenBank mRNA | AL833345.1 | - | 24 | 2461 | ||

TPTE
Genomic Map