siRNA Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005167.5 | NP_005158.5 | 10 | 1690 | ||
| GenBank mRNA | AK093270.1 | - | 10 | 1670 | ||
| AL133644.1 | - | 11 | 2454 | |||
| BC011803.2 | - | 10 | 1614 | |||
| BC064628.1 | - | 10 | 1472 | |||
| BC073828.1 | - | 10 | 1682 | 5244363 | ||

PPM1J
Genomic Map