siRNA Details
| Gene Symbol: CNKSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297647.1 | NP_001284576.1 | 21 | 2492 | ||
| NM_001297648.1 | NP_001284577.1 | 21 | 2466 | |||
| NM_006314.2 | NP_006305.2 | 21 | 2471 | |||
| XM_011540484.2 | XP_011538786.1 | 21 | 2451 | |||
| XM_011540485.2 | XP_011538787.1 | 18 | 1981 | |||
| GenBank mRNA | AB209335.1 | - | 21 | 2465 | ||
| AF100153.1 | - | 21 | 2449 | |||
| AK001900.1 | - | 21 | 2445 | |||
| AK222817.1 | - | 21 | 2450 | |||
| AK222904.1 | - | 21 | 2454 | |||
| AK292646.1 | - | 21 | 2453 | |||
| BC011604.2 | - | 21 | 2473 | 3139227 | ||
| BC012797.2 | - | 21 | 2473 | 3505293 | ||

CNKSR1
Genomic Map