siRNA Details
| Gene Symbol: LRRC41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006369.4 | NP_006360.3 | 10 | 2680 | ||
| GenBank mRNA | AF370430.1 | - | 10 | 2580 | ||
| AK125827.1 | - | 8 | 4191 | |||
| AK223420.1 | - | 7 | 2042 | |||
| AK291283.1 | - | 10 | 2663 | |||
| BC004953.1 | - | 7 | 1916 | |||
| BC092479.1 | - | 8 | 2045 | |||
| BX537366.1 | - | 10 | 2590 | |||
| X86018.1 | - | 7 | 1891 | |||

LRRC41
Genomic Map