siRNA Details
| Gene Symbol: LAPTM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006762.2 | NP_006753.1 | 8 | 1945 | ||
| XM_011542098.2 | XP_011540400.1 | 6 | 1778 | |||
| GenBank mRNA | D42042.1 | - | 8 | 1875 | ||
| U30498.1 | - | 8 | 1875 | |||
| U51240.1 | - | 8 | 1929 | |||

LAPTM5
Genomic Map