siRNA Details
| Gene Symbol: CFAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012337.2 | NP_036469.2 | 12 | 1643 | ||
| GenBank mRNA | AF094758.1 | - | 12 | 1675 | ||
| AK225566.1 | - | 12 | 1689 | |||
| AK225582.1 | - | 12 | 1689 | |||
| AK292249.1 | - | 12 | 1692 | |||
| BC027874.1 | - | 12 | 1643 | |||
| BC089391.1 | - | 12 | 1838 | 30387473 | ||
| BC109055.1 | - | 11 | 1658 | |||

CFAP45
Genomic Map