siRNA Details
Gene Symbol: SAC3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013299.3 | NP_037431.3 | 2 | 1389 | ||
GenBank mRNA | U79266.1 | - | 2 | 1384 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SAC3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013299.3 | NP_037431.3 | 2 | 1389 | ||
GenBank mRNA | U79266.1 | - | 2 | 1384 |