siRNA Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014313.3 | NP_055128.1 | 7 | 2062 | ||
| XM_005245817.1 | XP_005245874.1 | 6 | 1933 | |||
| XM_011541159.1 | XP_011539461.1 | 7 | 2256 | |||
| GenBank mRNA | AF081282.1 | - | 7 | 1990 | ||
| AL136627.1 | - | 7 | 2044 | |||
| AY071927.1 | - | 7 | 2039 | |||
| BC066908.1 | - | 7 | 2001 | 4817784 | ||
| BX537683.1 | - | 1 | 4986 | |||
| JX644905.1 | - | 7 | 2062 | |||
| JX644906.1 | - | 7 | 2062 | |||

TMEM50A
Genomic Map