siRNA Details
| Gene Symbol: CACYBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007214.1 | NP_001007215.1 | 6 | 1170 | ||
| NM_014412.2 | NP_055227.1 | 6 | 1355 | |||
| XM_017001046.1 | XP_016856535.1 | 6 | 1302 | |||
| GenBank mRNA | AF057356.1 | - | 6 | 1044 | ||
| AF314752.1 | - | 6 | 1054 | |||
| AK093425.1 | - | 6 | 1142 | |||
| AL035305.1 | - | 6 | 1047 | |||
| BC005975.1 | - | 6 | 1156 | 4102572 | ||
| BC022352.1 | - | 6 | 1053 | 4281339 | ||
| BC067823.1 | - | 6 | 1037 | 5260416 | ||
| BC078151.1 | - | 6 | 1020 | 6065872 | ||

CACYBP
Genomic Map