siRNA Details
| Gene Symbol: RBMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014469.4 | NP_055284.3 | 1 | 1613 | ||
| GenBank mRNA | AF069682.2 | - | 1 | 1519 | ||
| BC057796.1 | - | 1 | 1444 | 5261317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMXL2| Gene Symbol: RBMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014469.4 | NP_055284.3 | 1 | 1613 | ||
| GenBank mRNA | AF069682.2 | - | 1 | 1519 | ||
| BC057796.1 | - | 1 | 1444 | 5261317 | ||