siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 12, 13 | 2934 | ||
| XM_006711816.3 | XP_006711879.1 | 12, 13 | 2756 | |||
| XM_011544278.2 | XP_011542580.2 | 5, 6 | 545 | |||
| XM_017002391.1 | XP_016857880.1 | 11, 12 | 2498 | |||
| GenBank mRNA | AK021445.1 | - | 6, 7 | 910 | ||
| AK074078.1 | - | 7, 8 | 2039 | |||
| AK092353.1 | - | 6, 7 | 1330 | |||
| BC008300.1 | - | 7, 8 | 844 | |||

PCNX2
Genomic Map