siRNA Details
| Gene Symbol: RBM34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015014.2 | NP_055829.2 | 11 | 1185 | ||
| NR_027762.1 | 10 | 1048 | ||||
| XM_011544133.1 | XP_011542435.1 | 10 | 1485 | |||
| XM_017000721.1 | XP_016856210.1 | 11 | 1542 | |||
| GenBank mRNA | AK292362.1 | - | 11 | 1171 | ||
| BC029451.1 | - | 11 | 1185 | 4809263 | ||

RBM34
Genomic Map