siRNA Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015055.3 | NP_055870.2 | 3 | 492 | ||
| GenBank mRNA | AB014540.1 | - | 3 | 444 | ||
| AF134894.1 | - | 3 | 463 | |||
| AF210818.1 | - | 3 | 389 | |||
| AK304104.1 | - | 3 | 356 | |||
| BC000616.2 | - | 3 | 465 | 3346240 | ||
| DA413747.1 | - | 3 | 492 | |||

SWAP70
Genomic Map