siRNA Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297714.1 | NP_001284643.1 | 11 | 1742 | ||
| NM_015055.3 | NP_055870.2 | 12 | 1916 | |||
| XM_005252830.4 | XP_005252887.1 | 10 | 1817 | |||
| GenBank mRNA | AB014540.1 | - | 12 | 1868 | ||
| AF134894.1 | - | 12 | 1887 | |||
| AK027131.1 | - | 9 | 1343 | |||
| AK096859.1 | - | 11 | 1742 | |||
| AK304104.1 | - | 12 | 1780 | |||
| BC000616.2 | - | 12 | 1889 | 3346240 | ||

SWAP70
Genomic Map