siRNA Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136115.2 | NP_001129587.1 | 4 | 2688 | ||
| NM_015176.3 | NP_055991.1 | 5 | 2884 | |||
| NR_049764.1 | 4 | 2745 | ||||
| GenBank mRNA | AB007952.1 | - | 5 | 2635 | ||
| AK001628.1 | - | 5 | 2859 | |||
| BC031576.1 | - | 5 | 2851 | 5188219 | ||
| CN415382.1 | - | 1 | 325 | |||
| U38437.1 | - | 1 | 70 | |||

FBXO28
Genomic Map