siRNA Details
| Gene Symbol: SMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323614.1 | NP_001310543.1 | 6 | 800 | ||
| NM_001323615.1 | NP_001310544.1 | 6 | 800 | |||
| NM_001323616.1 | NP_001310545.1 | 7 | 932 | |||
| NM_001323617.1 | NP_001310546.1 | 7 | 767 | |||
| NM_015327.2 | NP_056142.2 | 6 | 739 | |||
| XM_017000842.1 | XP_016856331.1 | 2 | 685 | |||
| XM_017000843.1 | XP_016856332.1 | 6 | 746 | |||
| GenBank mRNA | AB029012.2 | - | 6 | 741 | ||
| AB085691.1 | - | 6 | 667 | |||
| AK092183.1 | - | 6 | 715 | |||
| AY168922.1 | - | 6 | 593 | |||
| AY336728.1 | - | 6 | 590 | |||
| BC038296.1 | - | 6 | 740 | 4824487 | ||

SMG5
Genomic Map