siRNA Details
| Gene Symbol: RASGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145648.1 | NP_001139120.1 | 25, 26 | 3991 | ||
| NM_002891.4 | NP_002882.3 | 26, 27 | 4039 | |||
| NM_153815.2 | NP_722522.1 | 12, 13 | 2485 | |||
| XM_017022455.1 | XP_016877944.1 | 26, 27 | 3930 | |||
| XM_017022456.1 | XP_016877945.1 | 25, 26 | 3900 | |||
| GenBank mRNA | AK226101.1 | - | 24, 25 | 3275 | ||
| L26584.1 | - | 26, 27 | 3670 | |||
| M91815.1 | - | 5, 6 | 532 | |||
| S62035.1 | - | 12, 13 | 2494 | |||

RASGRF1
Genomic Map