siRNA Details
| Gene Symbol: RBBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198719.1 | NP_001185648.1 | 3, 4 | 657 | ||
| NM_002893.3 | NP_002884.1 | 3, 4 | 669 | |||
| GenBank mRNA | AK091911.1 | - | 3, 4 | 654 | ||
| AK291567.1 | - | 3, 4 | 413 | |||
| BC114500.1 | - | 3, 4 | 567 | 40083852 | ||
| BC114501.1 | - | 3, 4 | 567 | 40083851 | ||
| CR541701.1 | - | 3, 4 | 311 | |||
| U35143.1 | - | 3, 4 | 572 | |||
| X72841.1 | - | 3, 4 | 596 | |||

RBBP7
Genomic Map