siRNA Details
| Gene Symbol: HINFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243259.1 | NP_001230188.1 | 10 | 2159 | ||
| NM_015517.4 | NP_056332.2 | 11 | 2214 | |||
| NM_198971.2 | NP_945322.1 | 10 | 2097 | |||
| XM_011542744.2 | XP_011541046.1 | 10 | 2206 | |||
| XM_011542745.2 | XP_011541047.1 | 10 | 2056 | |||
| XM_011542746.1 | XP_011541048.1 | 9 | 1877 | |||
| XM_017017499.1 | XP_016872988.1 | 10 | 2294 | |||
| XM_017017500.1 | XP_016872989.1 | 10 | 2173 | |||
| XM_017017501.1 | XP_016872990.1 | 11 | 2300 | |||
| XM_017017502.1 | XP_016872991.1 | 13 | 2485 | |||
| XM_017017503.1 | XP_016872992.1 | 11 | 2250 | |||
| XM_017017504.1 | XP_016872993.1 | 11 | 2125 | |||
| XM_017017505.1 | XP_016872994.1 | 11 | 2103 | |||
| XM_017017506.1 | XP_016872995.1 | 11 | 2066 | |||
| XM_017017507.1 | XP_016872996.1 | 13 | 2305 | |||
| XM_017017508.1 | XP_016872997.1 | 12 | 2180 | |||
| XM_017017509.1 | XP_016872998.1 | 10 | 1932 | |||
| GenBank mRNA | AK056362.1 | - | 10 | 2010 | ||
| AK092869.1 | - | 5 | 2351 | |||
| AL080201.1 | - | 2 | 907 | |||
| BC001073.2 | - | 10 | 2068 | 2820431 | ||
| BC001945.1 | - | 10 | 2068 | 2820431 | ||
| BC012856.1 | - | 10 | 2009 | 3871231 | ||
| BC017234.2 | - | 10 | 2017 | 3537381 | ||

HINFP
Genomic Map