siRNA Details
| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016002.2 | NP_057086.2 | 11 | 1499 | ||
| GenBank mRNA | AF151807.1 | - | 11 | 1149 | ||
| AK075178.1 | - | 11 | 1255 | |||
| BC026185.2 | - | 11 | 1160 | 4798194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCCPDH| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016002.2 | NP_057086.2 | 11 | 1499 | ||
| GenBank mRNA | AF151807.1 | - | 11 | 1149 | ||
| AK075178.1 | - | 11 | 1255 | |||
| BC026185.2 | - | 11 | 1160 | 4798194 | ||