siRNA Details
| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016002.2 | NP_057086.2 | 12 | 2321 | ||
| GenBank mRNA | AF151807.1 | - | 12 | 1969 | ||
| BM786495.1 | - | 1 | 567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCCPDH| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016002.2 | NP_057086.2 | 12 | 2321 | ||
| GenBank mRNA | AF151807.1 | - | 12 | 1969 | ||
| BM786495.1 | - | 1 | 567 | |||