siRNA Details
| Gene Symbol: PLEKHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304722.1 | NP_001291651.1 | 5 | 769 | ||
| NM_001304723.1 | NP_001291652.1 | 4 | 742 | |||
| NM_016274.5 | NP_057358.2 | 5 | 802 | |||
| XM_017001420.1 | XP_016856909.1 | 5 | 779 | |||
| GenBank mRNA | AF073836.1 | - | 4 | 432 | ||
| AF168676.1 | - | 5 | 534 | |||
| AF192912.1 | - | 5 | 739 | |||
| AF217956.1 | - | 5 | 590 | |||
| AF291105.1 | - | 5 | 730 | |||
| AK025937.1 | - | 3 | 219 | |||
| AK125609.1 | - | 3 | 2834 | |||
| AK222921.1 | - | 5 | 446 | |||
| AK302255.1 | - | 5 | 786 | |||
| BC010149.2 | - | 5 | 530 | 4330014 | ||
| BP329810.1 | - | 4 | 491 | |||
| BX353413.2 | - | 5 | 770 | |||
| BX376480.2 | - | 5 | 738 | |||
| BX394578.2 | - | 5 | 668 | |||

PLEKHO1
Genomic Map