siRNA Details
| Gene Symbol: PLEKHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304722.1 | NP_001291651.1 | 6 | 1261 | ||
| NM_001304723.1 | NP_001291652.1 | 5 | 1234 | |||
| NM_001304724.1 | NP_001291653.1 | 5 | 1159 | |||
| NM_016274.5 | NP_057358.2 | 6 | 1294 | |||
| XM_017001420.1 | XP_016856909.1 | 6 | 1271 | |||
| GenBank mRNA | AF073836.1 | - | 5 | 924 | ||
| AF168676.1 | - | 6 | 1026 | |||
| AF192912.1 | - | 6 | 1231 | |||
| AF217956.1 | - | 6 | 1082 | |||
| AF291105.1 | - | 6 | 1222 | |||
| AK025937.1 | - | 4 | 711 | |||
| AK125609.1 | - | 4 | 3326 | |||
| AK222921.1 | - | 6 | 938 | |||
| AK302255.1 | - | 6 | 1278 | |||
| BC010149.2 | - | 6 | 1022 | 4330014 | ||
| BC023533.2 | - | 5 | 1016 | |||

PLEKHO1
Genomic Map