siRNA Details
| Gene Symbol: TMEM138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016464.4 | NP_057548.1 | 4, 5 | 946 | ||
| NR_028473.1 | 5, 6 | 829 | ||||
| XM_006718585.3 | XP_006718648.1 | 4 | 1184 | |||
| XM_006718588.3 | XP_006718651.1 | 4, 5 | 441 | |||
| GenBank mRNA | AF151030.1 | - | 4, 5 | 563 | ||
| AF151032.1 | - | 4, 5 | 560 | |||
| AK027724.1 | - | 5 | 1095 | |||
| AL832879.1 | - | 3 | 3099 | |||
| BC005201.1 | - | 4, 5 | 561 | 3684390 | ||
| CB956757.1 | - | 5, 6 | 501 | |||

TMEM138
Genomic Map