siRNA Details
| Gene Symbol: ZCCHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282566.1 | NP_001269495.1 | 5 | 509 | ||
| NM_001282567.1 | NP_001269496.1 | 5 | 371 | |||
| NM_001282568.1 | NP_001269497.1 | 6 | 630 | |||
| NM_001282569.1 | NP_001269498.1 | 7 | 562 | |||
| NM_001282570.1 | NP_001269499.1 | 5 | 371 | |||
| NM_001282571.1 | NP_001269500.1 | 5 | 391 | |||
| NM_001282574.1 | NP_001269503.1 | 6 | 492 | |||
| NM_016505.3 | NP_057589.2 | 6 | 492 | |||
| XM_006710681.3 | XP_006710744.1 | 7 | 636 | |||
| XM_011541567.2 | XP_011539869.1 | 6 | 508 | |||
| GenBank mRNA | AF151077.1 | - | 6 | 421 | ||
| AF151085.1 | - | 6 | 448 | |||
| AF247661.2 | - | 5 | 361 | |||
| AK001929.1 | - | 6 | 442 | |||
| AK022506.1 | - | 6 | 479 | |||
| AK024049.1 | - | 6 | 468 | |||
| AK302248.1 | - | 7 | 512 | |||
| AK302343.1 | - | 5 | 317 | |||
| AK307478.1 | - | 5 | 345 | |||
| BC000685.2 | - | 6 | 530 | |||
| BC007446.2 | - | 6 | 428 | 3009823 | ||
| BC050609.1 | - | 6 | 432 | 6527025 | ||
| BP309837.1 | - | 5 | 332 | |||
| BP371153.1 | - | 5 | 327 | |||
| CN423139.1 | - | 6 | 429 | |||

ZCCHC17
Genomic Map