siRNA Details
| Gene Symbol: CEND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016564.3 | NP_057648.2 | 2 | 1378 | ||
| GenBank mRNA | AF235030.1 | - | 1 | 1284 | ||
| AK074547.1 | - | 2 | 1374 | |||
| BC034732.1 | - | 2 | 1364 | 5179740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEND1| Gene Symbol: CEND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016564.3 | NP_057648.2 | 2 | 1378 | ||
| GenBank mRNA | AF235030.1 | - | 1 | 1284 | ||
| AK074547.1 | - | 2 | 1374 | |||
| BC034732.1 | - | 2 | 1364 | 5179740 | ||